Organic reference standard, Furfural, analytical standard, Supelco®
04623-1ML
: Merck
:
- Förp.:1 mL
- Standard:Analytical standard
- CAS-nr:98-01-1
- Beskrivning:Furfural, ≥98,5% (GC)
- Smältpunkt:−36 °C (lit.)
- Formel:C₅H₄O₂
- Benämning:Furfural
- Kokpunkt:162 °C (lit.)
Furfural is a furan derivative obtained from renewable biomass-derived carbohydrates.
It is a sustainable alternative for petroleum-based building blocks used in the production of fine chemicals, plastics and more, also offers a promising rich platform for lignocellulosic biofuels.