Organic reference standard, DL-Menthol, analytical standard, Supelco®
Catalog # 05174-100MG
Supplier: Merck
CAS Number:
Specifications
- Förp.:100 mg
- Standard:Analytical standard
- CAS-nr:89-78-1
- Beskrivning:DL-Menthol, ≥98,0% (GC)
- Smältpunkt:34...36 °C (lit.)
- Formel:C₁₀H₂₀O
- Benämning:DL-Menthol
- Kokpunkt:216 °C (lit.)