Organic reference standard, Dimethyl succinate, analytical standard, Supelco®
Katalog # 73605-1ML
Leverantör: Merck
CAS-nummer:
Specifikationer
- Förp.:1 mL
- Standard:Analytical standard
- CAS-nr:106-65-0
- Beskrivning:Dimethyl succinate, ≥97,0% (GC)
- Smältpunkt:16...19 °C (lit.)
- Formel:C₆H₁₀O₄
- Benämning:Dimethyl succinate
- Kokpunkt:200 °C (lit.)
- Form:Neat