Organic reference standard, D-Camphor, analytical standard, Supelco®
50843-100MG
: Merck
:
- Förp.:100 mg
- Standard:Analytical standard
- CAS-nr:464-49-3
- Beskrivning:D-Camphor, ≥97,5% (GC)
- Smältpunkt:178...182 °C (lit.)
- Formel:C₁₀H₁₆O
- Benämning:D-Camphor
- Form:Neat
D-Camphor is an acyclic terpenoid that occurs naturally in the wood of Cinnamomum camphora.