Organic reference standard, Butyrophenone, analytical standard, Supelco®
16704-1ML
: Merck
:
- Förp.:1 mL
- Standard:Analytical standard
- CAS-nr:495-40-9
- Beskrivning:Butyrophenone, ≥99,0% (GC)
- Smältpunkt:11...13 °C (lit.)
- Formel:C₁₀H₁₂O
- Benämning:Butyrophenone
- Kokpunkt:228...230 °C (lit.)