Organic reference standard, Benzyl benzoate, analytical standard, Supelco®
Catalog # 68183-1ML
Supplier: Merck
CAS Number:
Specifications
- Förp.:1 mL
- Standard:Analytical standard
- CAS-nr:120-51-4
- Beskrivning:Benzyl benzoate, ≥98% (GC)
- Smältpunkt:17...20 °C (lit.)
- Formel:C₁₄H₁₂O₂
- Förvaringstemperatur:2...8 °C
- Benämning:Benzyl benzoate
- Kokpunkt:323...324 °C (lit.)
- Form:Neat
Specifications
About this item
Benzyl benzoate undergoes hydrogenation reaction catalyzed by a system of (Ru(acac)₃) (Where Ru-ruthenium/acac-acetylacetonate) with MeC(CH₂PPh₂)₃ (tripodal triphosphine ligand).
In the presence of excess of triethylamine in trifluoroethanol obtain benzyl alcohol.