Organic reference standard, Amyl salicylate, analytical standard, Supelco®
Katalog # 44041-250MG
Leverantör: Merck
CAS-nummer:
Specifikationer
- Förp.:250 mg
- Standard:Analytical standard
- CAS-nr:2050-08-0
- Beskrivning:Amyl salicylate, ≥98,0% (GC)
- Formel:C₁₂H₁₆O₃
- Benämning:Amyl salicylate