Organic reference standard, 2-Methyl-1-butanol, analytical standard, Supelco®
Catalog # 52566-1ML
Supplier: Merck
CAS Number:
Specifications
- Förp.:1 mL
- Standard:Analytical standard
- CAS-nr:3391-86-4
- Beskrivning:2-Methyl-1-butanol, ≥98,0% (GC)
- Formel:C₈H₁₆O
- Benämning:2-Methyl-1-butanol
- Form:Neat