Organic reference standard, 2-Methoxy-4-vinylphenol, analytical standard, Supelco®
Catalog # 89189-100MG
Supplier: Merck
CAS Number:
Specifications
- Förp.:100 mg
- Standard:Analytical standard
- CAS-nr:7786-61-0
- Beskrivning:2-Methoxy-4-vinylphenol, ≥98,0% (GC)
- Formel:C₉H₁₀O₂
- Förvaringstemperatur:2...8 °C
- Benämning:2-Methoxy-4-vinylphenol
- Flampunkt:113 °C
- Kokpunkt:224 °C (lit.)
- Form:Neat