ContactUsLinkComponent
Supelco® Organic Reference Standard, 1,4-Cineole
 :  Merck
undefined
Supelco® Organic Reference Standard, 1,4-Cineole
 :  Merck
  • CAS-nr:
    470-67-7
  • Storage conditions:
    2...30 °C
  • Faroklassbeskrivning:
    H226
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00209502
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    1-Isopropyl-4-methyl-7-oxabicyclo[2.2.1]heptane
  • Smältpunkt:
    –46 °C (lit.)
  • Formel:
    C₁₀H₁₈O
  • Molekylvikt:
    154,25
  • Densitet:
    0,887 g/ml at 25 °C (lit.)
  • Flampunkt:
    47 °C - closed cup
  • Kokpunkt:
    65 °C/16 mm Hg (lit.)
  • Benämning:
    1,4-Cineole