Order Entry
Sweden
ContactUsLinkComponent
Organic reference standard, 1,3,4,6,7,8,-Hexahydro-4,6,6,7,7,8-hexamethylcyclopenta[g]-2-benzopyran, analytical standard, Supelco®
Organic reference standard, 1,3,4,6,7,8,-Hexahydro-4,6,6,7,7,8-hexamethylcyclopenta[g]-2-benzopyran, analytical standard, Supelco®
Catalog # 57524-10MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, 1,3,4,6,7,8,-Hexahydro-4,6,6,7,7,8-hexamethylcyclopenta[g]-2-benzopyran, analytical standard, Supelco®
Catalog # 57524-10MG
Supplier:  Merck
CAS Number:  

Specifications

  • Förp.:
    10 mg
  • Standard:
    Analytical standard
  • CAS-nr:
    1222-05-5
  • Beskrivning:
    1,3,4,6,7,8,-Hexahydro-4,6,6,7,7,8-hexamethylcyclopenta[g]-2-benzopyran, ≥85,0% (GC)
  • Formel:
    C₁₈H₂₆O
  • Benämning:
    1,3,4,6,7,8,-Hexahydro-4,6,6,7,7,8-hexamethylcyclopenta[g]-2-benzopyran

Specifications

About this item