ContactUsLinkComponent
pH and conductivity in seawater, analytical standard, Supelco®
pH and conductivity in seawater, analytical standard, Supelco®
Katalog # PE3146-500ML
Leverantör:  Merck
CAS-nummer:  
pH and conductivity in seawater, analytical standard, Supelco®
Katalog # PE3146-500ML
Leverantör:  Merck
CAS-nummer:  
Specifikationer
  • Förp.:
    500 mL
  • Beskrivning:
    pH and Conductivity in Seawater