Pyridine-2,6-dicarboxylic acid 0.02 M i vatten eluent concentrate, for ion chromatography
Catalog # 50972-1L
Supplier: Merck
CAS Number:
Specifications
- Förp.:1 L
- Seq:5037
Specifications
About this item
:
:167.12 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:499-83-2
Formula:C₇H₅NO₄
MDL Number:MFCD00006299