ContactUsLinkComponent
Slide hybridisation oven, RapidFISH
Slide hybridisation oven, RapidFISH
Leverantör:  BOEKEL SCIENTIFIC
Certifikat
undefined
Slide hybridisation oven, RapidFISH
Leverantör:  BOEKEL SCIENTIFIC
Specifikationer
  • Catalog No:
  • BOEI240200-2EU
  • Modell:
  • RapidFISH
  • Display, upplösning:
  • Backlit 16×2 line blue LCD
  • Uppvärmningstid, RT till 65 °C:
  • 20 min
  • Operating mode:
  • Hold temperature, programmable temperature profile, calibration
  • Slide capacity:
  • 20 (25,4×76,2 mm) microscope slides
  • Larm:
  • Temp. deviation, step complete, program complete
  • Yttermått B×D×H:
  • 279,4×405,4×205,5 mm
  • Vikt:
  • 9,52 kg
Specifikationer