Organic acids kit, (21 components individually packaged), analytical standard, Supelco®
Catalog # 47264.
Supplier: Merck
CAS Number:
Specifications
- Koncentration:Neat
- Förp.:1
- Standard:Analytical standard
- Beskrivning:Organic Acids Kit
Specifications
About this item
Packaging: Storage temperature: –20°C