ContactUsLinkComponent
EPA 525 PAH mix B, certified reference material, Supelco®
EPA 525 PAH mix B, certified reference material, Supelco®
  48249.
 :  Merck
 :  
EPA 525 PAH mix B, certified reference material, Supelco®
  48249.
 :  Merck
 :  
  • Koncentration:
    500 mug/mL each component in acetone
  • Förp.:
    1
  • Beskrivning:
    EPA 525 PAH Mix B
  • Typ:
    Certified reference material