Dimethylpolysiloxane set of analytical standards, molecular weight series
Catalog # 361879-1SET
Supplier: Merck
CAS Number:
Specifications
- Förp.:1 SET
- Beskrivning:Dimethylpolysiloxane
set of analytical standards, molecular weight series
Specifications
About this item
Clear, colorless fluid polymer useful as a stationary phase in gas chromatography and as an anti-foaming agent.
It was used in a study performed to understand its molecular weight and molecular weight distribution by off-line coupling of GPC with MALDI mass spectrometer.
Kit components
Mw 40000
Mw 100000
Mw 270000
Certifications: Exact values can be taken from the accompanying certificate of analysis.