MacConkey agar, MP Biomedicals
Catalog # ICNA091006017
Supplier: MP Biomedicals
CAS Number:
Specifications
- Förp.:500 g
- Beskrivning:MacCONKEY agar
Specifications
About this item
A preparation for the isolation of enteric bacilli based on the fermentation of lactose.
Ready for CE IVD labeling of clinical applications.
Caution: Research use only (RUO).