Screw cap, DURAN® PURE GLS 80®
Katalog # SCOT891169109
Leverantör: DWK Life Sciences
Specifikationer
- Ø:88 mm
- Gänga:GLS 80
- Färg:Vit
- Förp.:1
Specifikationer
Om denna produkt
The DURAN® PURE GLS 80® screw cap is made from PSU with highly inert PTFE coated silicone seal. It is designed for use with DURAN® PURE GLS 80® bottles.
- Compatible with autoclaving and other common sterilization methods
- With batch identification moulded into cap
- Packaged in ISO Class 7 clean room
- Single batch delivery
- Certificate of Analysis and Change Control on request
Typical applications: Manufacturing of pharmaceutical products, storage of bulk drug intermediates and ingredients, transportation of APIs.