Micro spatula, stainless steel
BURK5386-0001
: BURKLE
- Kapacitet:1,25 ml
- Längd:170 mm
- Förp.:1
The micro spatula can be used for reliably measuring, sampling, or processing small amounts of powder, chemicals, pastes, creams, or active ingredients.
- Stainless steel AISI 304 (1.4301)
- High-gloss polished
The laboratory scoop is manufactured entirely without edges, crevices, or undercuts. This eliminates the possibility of impurities or cross-contamination, as no product residue or cleaning agents can be deposited. The micro spatula is ideal for use in the pharmaceutical industry as well as food and beverage, and cosmetics.