Tamper-proof sample bags, SteriBag Premium
Catalog # BURK5344-8000
Supplier: BURKLE
Specifications
- Kapacitet:300 ml
- L×B:200×145 mm
- Förp.:100
Specifications
About this item
The SteriBag Premium sample bags uniquely combine a high degree of security, sterility, tamper evident closure and traceability, they are used to securely transport and for the storage of samples.
- Sterilised by gamma rays (SAL 10⁻⁶)
- With integrated tamper-proof closure for manipulation evidence
- Unique identification via tracking number
- Suitable for the use with foodstuffs
After sampling, the sample can be filled directly into the sterile sample bag. The tamper-evident closure seals the bag liquid-tight and secure. When trying to open the closure again, the word 'STOP' appears clearly visible. Both the sample bag and the tear-off field are labeled with a unique tracking number, this allows a complete documentation and traceability.
Suitable for many demanding applications in the food, pharmaceutical, medical area as well as in cleanroom or biotechnology sectors.