Macron Fine Chemicals™ StandARd®, Platinum Cobalt Colour Standard (APHA 500)
Katalog # H296-03
Leverantör: MACRON AVANTOR BRAND
CAS-nummer:
Specifikationer
- Förpackningstyp:Glass bottle, amber, narrow mouth
- Förp.:500 mL
- Beskrivning:Platinum Cobalt (Pt-Co) colour standard, APHA 500
- Seq:02
Specifikationer
Om denna produkt
Corrosive.
APHA 500 contains 0,13% chloroplatinic acid, 0,11% cobalt (II) chloride, and 4,6% hydrogen chloride in water.