FLED fluorescence intermediate attachments
Leverantör: MOTIC
Gå till:
Om denna produkt
Compatible with Panthera U, KU and CC microscopes.
3 Alternativ tillgängliga nedan
Product Details & Documents
Compatible with Panthera U, KU and CC microscopes.
Product Family Options
Produktinformation
- TillgänglighetPrice
- MOTI1101011400181FLED Fluorescence intermediate attachment (Auramine O)
- Fluorescence intermediate part
- LED excitation module AO, consisting of 455 nm LED lamp and AO filter cube
- Light blocking slider
Note: Recommended to use with Plan objectives UC PL NCGFLED Fluorescence intermediate attachment (Auramine O)
- Fluorescence intermediate part
- LED excitation module AO, consisting of 455 nm LED lamp and AO filter cube
- Light blocking slider
Note: Recommended to use with Plan objectives UC PL NCGLog in för att kolla tillgänglighet och ert pris1 * 1 Items24 400,00 kr - MOTI1101011400191FLED Fluorescence intermediate attachment (FITC)
- Fluorescence intermediate part
- LED excitation module FITC, consisting of 470 nm LED lamp and FITC filter cube
- Light blocking sliderFLED Fluorescence intermediate attachment (FITC)
- Fluorescence intermediate part
- LED excitation module FITC, consisting of 470 nm LED lamp and FITC filter cube
- Light blocking sliderLog in för att kolla tillgänglighet och ert pris1 * 1 Items21 275,00 kr - MOTI1101011400201FLED Fluorescence intermediate attachment (TRITC)
- Fluorescence intermediate part
- LED excitation module TRITC, consisting of 525 nm LED lamp and TRITC filter cube
- Light blocking sliderFLED Fluorescence intermediate attachment (TRITC)
- Fluorescence intermediate part
- LED excitation module TRITC, consisting of 525 nm LED lamp and TRITC filter cube
- Light blocking sliderLog in för att kolla tillgänglighet och ert pris1 * 1 Items21 275,00 kr