Digital Solutions
Applications
Support
About
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
Sweden
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
ContactUsLinkComponent
Hem
Standarder
Petroleum Industry Standards
WIP VOA standard, certified reference material, Supelco®
WIP VOA standard, certified reference material, Supelco®
WIP VOA standard, certified reference material, Supelco®
Catalog # UST104-1.5ML
Supplier:
Merck
CAS Number:
Expand image
WIP VOA standard, certified reference material, Supelco®
Catalog # UST104-1.5ML
Supplier:
Merck
CAS Number:
Specifications
Förp.:
1,5 mL
Beskrivning:
WIP VOA standard
Specifications
About this item
Product Details
Product Details