Bossheads
Katalog # RLAAZ0204
Leverantör: R L ENTERPRISES
Specifikationer
- Beskrivning:Bosshead with blue plastic headed screw
- Material:Gjuten legering
- Förp.:1
- För stavdiameter:16 mm
Specifikationer
Om denna produkt
Bossheads made from nickel plated cast alloy.
- With blue plastic headed nickel plated steel screws
- For use with rods up to 16 mm