Desiccant pack, Whatman™
Catalog # WHAT10548239
Supplier: Whatman products (Cytiva)
Specifications
- Beskrivning:Desiccant pack
- Storlek:1 g
- Förp.:100
Specifications
About this item
Desiccant pack ensure that DNA sampling cards remain dry during transport or storage.
- High vapour transmission rate
- Substrate is made with Tyvek® material
- Non-dusting, strong and hard to tear