Digital Solutions
Applications
Support
About
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
Sweden
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
ContactUsLinkComponent
Hem
Standarder
Standarder för miljöanalys
Trace Metals 2-WP, (CRM), Supelco®
Trace Metals 2-WP, (CRM), Supelco®
Trace Metals 2-WP, (CRM), Supelco®
Catalog # QC1052-20ML
Supplier:
Merck
CAS Number:
Expand image
Trace Metals 2-WP, (CRM), Supelco®
Catalog # QC1052-20ML
Supplier:
Merck
CAS Number:
Specifications
Förp.:
20 mL
Beskrivning:
Trace Metals 2-WP
Typ:
Certified reference material
Specifications
About this item
Product Details
Product Details