Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standarder
Standarder för miljöanalys
Lead free paint - powdered, (CRM), Supelco®
Lead free paint - powdered, (CRM), Supelco®
Lead free paint - powdered, (CRM), Supelco®
Katalog # CRM050-20G
Leverantör:
Merck
CAS-nummer:
Expandera bilden
Lead free paint - powdered, (CRM), Supelco®
Katalog # CRM050-20G
Leverantör:
Merck
CAS-nummer:
Specifikationer
Förp.:
20 g
Beskrivning:
Lead Free Paint - Powdered
Pk Info:
Pkg of 20 g
Typ:
Certified reference material