Supelco® TraceCERT® Organic Reference Standard, Amino Acids Mix Solution
Katalog # 79248-5X2ML
Leverantör: Merck
CAS-nummer:
Specifikationer
- Förp.:5
- Standard:Certified reference material, TraceCERT®
- Beskrivning:Amino acids mix solution
- Storage conditions:–20 °C
- Faroklassbeskrivning:H290
- Shipping conditions:STD
- M-Clarity quality level:MQ300
Specifikationer
Om denna produkt
- GHS signal: GHS05
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Caution: P234 - P390