Digital Solutions
Applications
Support
About
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
Sweden
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
ContactUsLinkComponent
Hem
Standarder
Organic Standards
Pharmaceutical secondary standard, methylparaben, (CRM), Supelco®
Pharmaceutical secondary standard, methylparaben, (CRM), Supelco®
Pharmaceutical secondary standard, methylparaben, (CRM), Supelco®
Leverantör:
Merck
Certifikat
Expandera bilden
Pharmaceutical secondary standard, methylparaben, (CRM), Supelco®
Leverantör:
Merck
Certifikat
Om denna produkt
Specifikationer
CAS-nr:
99-76-3
Benämning:
Methylparaben
Specifikationer
Relaterad information
Dokumentation
Certifikat
Sök efter LOT-nummer
Produktlitteratur
Merck Pharma Segment Brochure
Dokumentation