Supelco® TraceCERT® Organic Reference Standard, Triclosan
Katalog # 93453-100MG
Leverantör: Merck
CAS-nummer:
Specifikationer
- Förp.:100 mg
- Standard:Certified reference material, TraceCERT®
- CAS-nr:3380-34-5
- Beskrivning:Triclosan
- Storage conditions:15...25 °C
- Faroklassbeskrivning:H315 - H319 - H410
- Shipping conditions:STD
- MDL No.:MFCD00800992
- M-Clarity quality level:MQ300
- Synonym(s):5-Chloro-2-(2,4-dichlorophenoxy)phenol; Irgasan
- Molekylvikt:289,54
Specifikationer
Om denna produkt
- GHS signal: GHS07, GHS09
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Caution: P264 - P273 - P280 - P302 + P352 - P305 + P351 + P338 - P332 + P313