Order Entry
Sweden
Orders LinkContactUsLinkComponent
 

Supelco® TraceCERT® Organic Reference Standard, Glyphosate

Glyphosate, Certified reference material, TraceCERT®, 100 mg
Catalog # 89432-100MG
Supplier:   Merck
Supelco® TraceCERT® Organic Reference Standard, Glyphosate
Catalog # 89432-100MG
Supplier:  Merck
CAS Number:  
Supelco® TraceCERT® Organic Reference Standard, Glyphosate
Supelco® TraceCERT® Organic Reference Standard, Glyphosate

Glyphosate, Certified reference material, TraceCERT®, 100 mg

CAS Number:  
Price
843,00 kr
Log in for availability
1 * 100 mg
Product Details & Documents
  • GHS signal: GHS05, GHS09

Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.

Caution: P273 - P280 - P302 + P352 + P312 - P305 + P351 + P338 - P362 + P364 - P391

Specifications

Specifications

  • Förp.:
    100 mg
  • Standard:
    Certified reference material, TraceCERT®
  • Beskrivning:
    Glyphosate
  • Förvaringsförhållanden:
    15...25 °C
  • Faroklassbeskrivning:
    H312 - H318 - H411
  • Cat. No.:
    89432-100MG
  • M-Clarity quality level:
    MQ300
  • CAS-nr:
    1071-83-6
  • Smältpunkt:
    230 °C (dec.) (lit.)
  • MDL No.:
    MFCD00055350
  • Molekylvikt:
    169,07
  • Synonym(s):
    N-(Phosphonomethyl)glycine; Glyphosate
  • Shipping conditions:
    STD
  • Formel:
    (HO)₂P(O)CH₂NHCH₂CO₂H

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.