Flat Bags
Katalog # KWPA123045
Leverantör: KWP KUNSTSTOFF WERK PLUR
Specifikationer
- Färg:Genomskinlig
- Beskrivning:Flat bag, transparent
- Mått:200×240 mm
- Tjocklek:0,05 mm
- Förp.:1.000
Specifikationer
Om denna produkt
Flat bag made from LDPE.
- Suitable for cleanroom applications (ISO class 5)
- LDPE grade for pharmaceutical applications