Lux kit, Testo 440
Katalog # TEST0563.4402
Supplier: TESTO
Specifications
- Beskrivning:Air velocity and IAQ measuring instrument
- Seq:72
- Förp.:1
Specifications
About this item
Lux kit is equipped with clearly structured measurement menu for long-term measurement and determination of illuminance according to the V-lambda curve , thus for all common light sources.
- Designed for measuring the illuminance in workplaces
- Turbulence measurement in accordance with EN ISO 7730 / ASHRAE 55
Non-slip probe contact surface for convenient positioning at the measuring location, internal data storage and USB port for data export. The air velocity and IAQ measuring instrument can be expanded with a large portfolio of digital probes.
Ideal for different range of probes for cleanrooms and laboratories, long-term monitoring of indoor air quality, convenience and maximum flexibility for duct and outlet measurements.
Certifications: Standard-compliant accuracy according to DIN EN 13032-1 and class C according to DIN 5032-7.
Delivery: Supplied with testo 440 air velocity and IAQ measuring instrument, 3 x AA batteries, USB cable and calibration protocol (TEST0560.4401), Lux probe with fixed cable (cable length 1,4 m) and calibration protocol (TEST0635.0551), basic case for testo 440 and one probe.