Sterilisation indicator tapes, Stericlin®
Product Details & Documents
These adhesive tapes are ideal for safe and sterilisation-proof sealing of all curved packaging.
- For steam sterilisation
- Beige tapes are intended for use with crepe sheet packaging
- Blue tapes are for use with non woven and SMS sheet packaging
Available with or without indicator print.
: Please note: Do not stick to metal or plastic containers - adhesive residues may remain behind.
Specifikationer
- Bredd:25 mm
- Beskrivning:Adhesive tapes with STEAM indicator, beige
- Längd:50 m
- Seq:03
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.