Blades for safety knives, No. 86, Styropor
Katalog # MARO86.60
Leverantör: MARTOR ARGENTAX
Specifikationer
- Beskrivning:Replacement blade, No. 86 Styropor, double bevel edge-ground, 2-sided ground-edge
- L×B:95×8,90 mm
- Tjocklek:0,40 mm
- Förp.:10
Specifikationer
Om denna produkt
Styropor blade for use with safety knives.
- 2-sided ground-edge
- Double bevel edge-ground
Mountable in:
ARGENTAX CUTTEX