Universal vials
Universal vials, Nej
Catalog # EKSIA62251
Supplier: EKSIGENT
Jump to:
Product Details & Documents
Universal vials are used to accommodate buffers/samples.
These polymethylpentene vials are precision manufactured and have been compatibility tested with commonly used CE reagents. Universal vials are designed for use only with CESI 8000, PA 800 Enhanced or PA 800 plus instruments and should not be reused.
Specifications
- Med lock:Nej
- Beskrivning:Universal vials