Chemical resistant gloves, u-chem 3300
Leverantör: UVEX
Certifikat
Om denna produkt
These highly sensitive chemical protection gloves with gauntlet, made of seamless bamboo-viscose fibre and a nitrile rubber coating, are kind to skin and comfortable to wear even for long periods.
- Extremely lightweight and highly flexible
- Very good fit
- Good resistance to many chemicals
- Silicone-free according to imprint test
- Low-linting
Ideal for use in chemical industry, printing industry, maintenance and inspection work, laboratory tasks, painting work, food processing, pharmaceutical industry and in cleaning.
Certifications: EN 388:2016 (2121X), EN ISO 374-1:2016/Type A (JKLOPT).
Specifikationer
- Catalog No:
- 112-5412
- 112-5413
- 112-5416
- Modell:
- 3300
- 3300
- 3300
- Ej handspecifika/handspecifika:
- Handspecifika
- Handspecifika
- Handspecifika
- Engångs/Återanvändbar:
- Återanvändbar
- Återanvändbar
- Återanvändbar
- Latex/latexfri:
- Latexfria
- Latexfria
- Latexfria
- Pudrad/puderfri:
- Puderfria
- Puderfria
- Puderfria
- Fodermaterial:
- Nylon
- Nylon
- Nylon
- Yta:
- Fully coated
- Fully coated
- Fully coated
- Kragmodell:
- Kraghandske
- Kraghandske
- Kraghandske
- EN standard:
- EN 388:2016 (2121X), EN ISO 374-1:2016 Type A (JKLOPT)
- EN 388:2016 (2121X), EN ISO 374-1:2016 Type A (JKLOPT)
- EN 388:2016 (2121X), EN ISO 374-1:2016 Type A (JKLOPT)
- Godkända för livsmedelskontakt:
- +
- +
- +