UV exposure screen for myECL™ imager, Pierce
Catalog # PIER62244
Supplier: Thermo Fisher Scientific
Specifications
- Beskrivning:UV exposure screen for myECL™ imager, 245×195×3,2 mm
- Förp.:1
Specifications
About this item
This clear plastic sample screen is used to cover the transilluminator on the myECL™ imager. It is a replacement for the accessory used on the transilluminator for acquiring images in the UV mode.