ContactUsLinkComponent
Chemiluminescence exposure screen for myECL™ imager
Chemiluminescence exposure screen for myECL™ imager
Catalog # PIER62242
Chemiluminescence exposure screen for myECL™ imager
Catalog # PIER62242
Specifications
  • Beskrivning:
    Chemiluminescence exposure screen for myECL™ imager, 245×195×3,2 mm
  • Förp.:
    1
Specifications
About this item
This black plastic sample screen is used to cover the transilluminator on the myECL™ imager.
This has been used as a replacement for the accessory used on the transilluminator for acquiring chemiluminescent images.