Liquid acid cleaner, TASKI Sani Cid
Catalog # DIVY7512833
Supplier: Diversey
Specifications
- Kapacitet:6×1 l
- Beskrivning:Sani Cid, liquid detergent
- Environmentally Preferable:Safer Human and Environmental Health,Sustainable Materials
- Miljöcertificering:EU Ecolabel DK/020/006
- Förp.:6 Bottle
Specifications
About this item
This is a mildly acidic liquid cleaner that is ideal for removing soap scum and hard water deposits from most washroom surfaces.
- Citric acid base is safe to use on all acid resistant surfaces including chrome and stainless steel
- Pleasant ONT fragrance
Streak-free drying allows staff to clean surfaces with the same product used for showers, doors, porcelain, counter tops, sinks and stainless steel.