Orders LinkContactUsLinkComponent
Mini LED light box
Mini LED light box
Mini-Light Box, 161×135×44 mm, Mini-magnifier
Katalog # HEAT120661
Leverantör:   HEATHROW
Mini LED light box
Katalog # HEAT120661
Leverantör:  HEATHROW
Mini LED light box
Mini LED light box
Mini-Light Box, 161×135×44 mm, Mini-magnifier
Price
3 600,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1 Items
Produktdetaljer och dokument
This small light box saves bench space and can be stored in a drawer.
  • Slim light panel
  • LED light source stays cool and has longer life (10000 hours)
  • Low power consumption
Viewing area 125×100 mm is perfect for quick screening of 96-well plates, Petri dishes, electrophoresis gels, slides, and transparencies.
Leveransinformation: Includes six 1.5 V AAA batteries.
Specifikationer
  • Beskrivning:
    Mini-Light Box, 161×135×44 mm
  • För:
    Mini-magnifier