USP calibration mix #7
Katalog # REST36009
Leverantör: Restek
CAS-nummer:
Specifikationer
- Koncentration:In dimethyl sulphoxide
- Förp.:1
- Beskrivning:USP <467> calibration mixture #7
- Pk Info:1 ml/ampoule
Specifikationer
Om denna produkt
The mix consists of chloroform (60 μg/ml), 1,4-dioxane (380 μg/ml), methylene chloride (dichloromethane) (600 μg/ml) and trichloroethene (80 μg/ml).
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.