Cleanroom wipes, Clino® one way
Catalog # PPSA3600070
Supplier: PFENNIG REINIGUNGSTECHNIK
Specifications
- Färg:Vit
- L×B:230×230 mm
- Förp.:300
Specifications
About this item
Clino® one way clean surfaces in semiconductors and pharma industries.
- More excellent intake capacity
- High abrasion resistance
- Low particle and fibre emission
Suitable for use in ISO class 7 and 8 (GMP grade C and D) cleanrooms.