Diffuser caps for waste and wash vials
Katalog # PERCN6101276
Leverantör: PerkinElmer
Specifikationer
- Beskrivning:Vial diffuser for waste and wash vials
- Förp.:1
Specifikationer
Om denna produkt
The conical shape allows the needle to penetrate in to solvent or waste.
- Reduce spillage and large losses through evaporation