Softshell jackets, WX3
Om denna produkt
Portwest WX3 contemporary workwear is designed to the highest standards and offer a solution for everyday activity and working environments. Constructed with cutting edge fabrics giving superior abrasion resistance and exceptional all-day comfort. The softshell jacket features a windproof, water-resistant and breathable membrane with a micro polar fleece.
- Chin guard for added comfort and stability
- Back vent for breathability and comfort
- Pre-bent sleeves allow for increased freedom of movement
- Elastic bound hem and cuffs for comfort and warmth
- Ezee zip allows for quick and easy fastening even when wearing work gloves
- 9 pockets for ample storage
- Sleeve pocket
- Zipped chest pocket
Weight: 310 g (94% polyester, 6% elastane 160 g laminated to 100% polyester micro polar fleece 150 g)
6 Alternativ tillgängliga nedan
Produktdetaljer och dokument
Portwest WX3 contemporary workwear is designed to the highest standards and offer a solution for everyday activity and working environments. Constructed with cutting edge fabrics giving superior abrasion resistance and exceptional all-day comfort. The softshell jacket features a windproof, water-resistant and breathable membrane with a micro polar fleece.
- Chin guard for added comfort and stability
- Back vent for breathability and comfort
- Pre-bent sleeves allow for increased freedom of movement
- Elastic bound hem and cuffs for comfort and warmth
- Ezee zip allows for quick and easy fastening even when wearing work gloves
- 9 pockets for ample storage
- Sleeve pocket
- Zipped chest pocket
Weight: 310 g (94% polyester, 6% elastane 160 g laminated to 100% polyester micro polar fleece 150 g)
Dokument
TDS T750 WX3 Softshell Jacket
Datablad
Specifications for Product Family
Specifikationer
- Catalog No:
- 113-9403
- 113-9402
- 113-9401
- 113-9404
- 113-9405
- 113-9406
- Modell:
- WX3
- WX3
- WX3
- WX3
- WX3
- WX3
- Outfit applikationer:
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- Risk area:
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Kön:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Stängningsstorlek:
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Tyg:
- 94% polyester, 6% elastane 160 g laminated to 100% polyester micro polar fleece
- 94% polyester, 6% elastane 160 g laminated to 100% polyester micro polar fleece
- 94% polyester, 6% elastane 160 g laminated to 100% polyester micro polar fleece
- 94% polyester, 6% elastane 160 g laminated to 100% polyester micro polar fleece
- 94% polyester, 6% elastane 160 g laminated to 100% polyester micro polar fleece
- 94% polyester, 6% elastane 160 g laminated to 100% polyester micro polar fleece
- Vikt:
- 310 g/m²
- 310 g/m²
- 310 g/m²
- 310 g/m²
- 310 g/m²
- 310 g/m²
Specifikationer
Product Family Options
Produktinformation
- FärgKlädstorlekTillgänglighetPrice
- Specifikationer:
More
Specifications
Cat. No.113-9403ModellWX3Outfit applikationerGeneral PurposeRisk areaRain (waterproof)KönUnisexSleeve or leg lengthLongStängningsstorlekMed dragkedjaTyg94% polyester, 6% elastane 160 g laminated to 100% polyester micro polar fleeceVikt310 g/m² - Specifikationer:
More
Specifications
Cat. No.113-9402ModellWX3Outfit applikationerGeneral PurposeRisk areaRain (waterproof)KönUnisexSleeve or leg lengthLongStängningsstorlekMed dragkedjaTyg94% polyester, 6% elastane 160 g laminated to 100% polyester micro polar fleeceVikt310 g/m² - Specifikationer:
More
Specifications
Cat. No.113-9401ModellWX3Outfit applikationerGeneral PurposeRisk areaRain (waterproof)KönUnisexSleeve or leg lengthLongStängningsstorlekMed dragkedjaTyg94% polyester, 6% elastane 160 g laminated to 100% polyester micro polar fleeceVikt310 g/m² - Specifikationer:
More
Specifications
Cat. No.113-9404ModellWX3Outfit applikationerGeneral PurposeRisk areaRain (waterproof)KönUnisexSleeve or leg lengthLongStängningsstorlekMed dragkedjaTyg94% polyester, 6% elastane 160 g laminated to 100% polyester micro polar fleeceVikt310 g/m² - Specifikationer:
More
Specifications
Cat. No.113-9405ModellWX3Outfit applikationerGeneral PurposeRisk areaRain (waterproof)KönUnisexSleeve or leg lengthLongStängningsstorlekMed dragkedjaTyg94% polyester, 6% elastane 160 g laminated to 100% polyester micro polar fleeceVikt310 g/m² - Specifikationer:
More
Specifications
Cat. No.113-9406ModellWX3Outfit applikationerGeneral PurposeRisk areaRain (waterproof)KönUnisexSleeve or leg lengthLongStängningsstorlekMed dragkedjaTyg94% polyester, 6% elastane 160 g laminated to 100% polyester micro polar fleeceVikt310 g/m²