Oxidase sticks for microbiology
Catalog # APPC416444.2326
Supplier: PanReac AppliChem
CAS Number:
Specifications
- Förp.:50
- Beskrivning:Oxidase sticks for microbiology
Specifications
About this item
Plastic sticks used for the easy and rapid detection of the cytochrome oxidase enzyme in microbiological diagnosis. Oxidase sticks have a much higher stability compared to the traditional laboratory test, because the tetramethyl-p-phenylenediamine reagent is attached to the pad.
Caution: *Produkten/produkterna på denna sida ligger utanför tillämpningsområdet för EU -direktivet 2019/904