Standard ground joint, bent
Catalog # QVFLGSG0108003_D11
Supplier: QVF LABORTECHNIK
Specifications
- Kärna:29/32 NS
- Beskrivning:Joint with standard ground joint cone
- Förp.:1
Specifications
About this item
The bent ground joint is fitted with glass thread GL 14.