DNA transfection, LipoMag™ transfection kit
Leverantör: OZ BIOSCIENCES
Certifikat
Om denna produkt
LipoMag™ transfection kit combines the efficiency of DreamFect™ Gold with CombiMag™ magnetic nanoparticles. By taking advantage of Magnetofection transfection techniques, the LipoMag™ kit reaches even higher transfection efficiency, especially in primary cells, while reducing toxicity. It is suitable for all type of nucleic acids such as plasmid DNA, linearised DNA, siRNA, oligonucleotides, double stranded RNA, mRNA and shRNA.
- Highest efficiency without toxicity
- Universal (cell lines and primary cells)
- Simple, ready to use and rapid
- Serum compatible
Accessories information: This kit needs to be used with a magnetic plate.
Delivery: Shipped at room temperature.
Caution: Store DreamFect™ Gold –20 °C and CombiMag™ at +4 °C.
Specifikationer
- Cellspecifik:Hard-to-transfect