Ring light, VisiLED Slim
VisiLED slim ring light, FWD 40 - 90 mm and 20 - 40 mm, int.Ø 66 mm, ext.Ø 82 mm, max. 65 klx (at 50 mm FWD) and 45 klx (at 25 mm FWD)
Katalog # SCOC400150
Leverantör: SCHOTT AG Lighting and imaging
Gå till:
Product Details & Documents
The VisiLED Slim is a space saving LED ring light, optimal for unconfined object preparation or usage with objective revolvers.
- Easily adaptable to microscope objectives by means of thumb screw
- Homogeneous incident or oblique illumination
- Controllable with all VisiLED controllers (MC750, MC1000, MC1500)
- Variable working distance ranges possible by exchange of focus optic rings
- Implementation of brightfield and darkfield applications
- Segments controllable in different preset modes
Documents
Ring Lights, EasyLED
Videor
TDS VisiLED Slim Ringlight
Datablad
VisiLED Series Professional Illumination for Stereo Microscopy
Produktlitteratur
Specifikationer
- Beskrivning:VisiLED slim ring light, FWD 40 - 90 mm and 20 - 40 mm, int.Ø 66 mm, ext.Ø 82 mm, max. 65 klx (at 50 mm FWD) and 45 klx (at 25 mm FWD)
- Seq:647